pH buffers, DuraCal
4,01, ±0,02, Pink, Plastic bottle, 3×500 ml, 3
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
For full information concerning health and safety data please contact Avantor or Hamilton directly.
- Label includes expiration date, batch number and actual value
- Traceable to NIST and PTB, certificates available to download
- Stable solutions
- Certified pH value from a DAkkS laboratory accredited for pH measurement
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:3
- Colour:Pink
- pH accuracy:±0,02
- Pk Info:3×500 ml
- pH value:4,01



