pH buffers, DuraCal
10,01, ±0,02, Carbonate, Yellow, Plastic bottle, 500 ml
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
For full information concerning health and safety data please contact Avantor or Hamilton directly.
- Label includes expiration date, batch number and actual value
- Traceable to NIST and PTB, certificates available to download
- Stable solutions
- Certified pH value from a DAkkS laboratory accredited for pH measurement
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Carbonate
- Pk:500 ml
- Colour:Yellow
- pH accuracy:±0,02
- pH value:10,01



