Order Entry
Export
Orders LinkContactUsLinkComponent
Phosphate standard solutions

Phosphate standard solutions

Phosphate standard solution, 500 ml (NIST), 1 mg/l as PO43-, 500 mL
Catalog # HACH256949
Supplier:   Hach
Phosphate standard solutions
Catalog # HACH256949
Supplier:  Hach
CAS Number:  
Phosphate standard solutions
Phosphate standard solutions

Phosphate standard solution, 500 ml (NIST), 1 mg/l as PO43-, 500 mL

CAS Number:  
Price
41,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solution for accuracy checks in phosphate (phosphorus) determinations.

Specifications

Specifications

  • Concentration:
    1 mg/l as PO<sub>4</sub><sup>3-</sup>
  • Pk:
    500 mL
  • Description:
    Phosphate standard solution, 500 ml (NIST)

Specifications