Nessler reagent
500 mL
Supplier: Hach
Jump to:
Product Details & Documents
The reagent is used for ammonia nitrogen determination in waste water by the Nessler method
Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
- CAS Number:7783-33-7
- MDL Number:MFCD00049656
Specifications
Specification Test Results
- Range:0.02-2.50 mg/L NH3-N
Specifications
- Pk:500 mL



