Aluminium standard solution
Aluminium, Al, 100 mg/L, 100 mL
Supplier: Hach
Jump to:
Product Details & Documents
Standard solution for accuracy checks in aluminium determinations.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:100 mg/L
- Pk:100 mL
- Element:Aluminium
- Symbol:Al