Orders LinkContactUsLinkComponent
Accessories for Optima™ heated circulating baths
Accessories for Optima™ heated circulating baths
Stainless steel VR test tube rack for 16-19 mm Ø tubes, 36 tubes
Catalog # 462-8258
Supplier:   GRANT INSTRUMENTS
Accessories for Optima™ heated circulating baths
Catalog # 462-8258
Supplier:  GRANT INSTRUMENTS
Accessories for Optima™ heated circulating baths
Accessories for Optima™ heated circulating baths
Stainless steel VR test tube rack for 16-19 mm Ø tubes, 36 tubes
Price
144,00 €
Log in for availability
1 * 1 items
Product Details & Documents
A range of different stainless steel accessories to use with Optima™ baths.
  • Raised shelves help accommodate different sizes of vessel by altering the effective liquid depth
  • Robust test tube racks are available in a variety of configurations for all sizes of tubes
Ordering information: ST5 and P5 baths can accommodate 1 of the following racks: 462-8251, 462-8252, 462-8253, 462-8254, 462-8255 and 462-8256, the ST12 or P12 and larger baths can accommodate 2 or more of the following racks 462-8257, 462-8258, 462-8133, 462-8134, 462-8261 or 462-8262.
Specifications
  • Description:
    Stainless steel VR test tube rack for 16-19 mm Ø tubes
  • Seq:
    10
  • For:
    36 tubes