Accessories for Optima™ heated circulating baths
Raised shelf, ST18/P18
Catalog # 462-8264
Supplier: GRANT INSTRUMENTS
Jump to:
Product Details & Documents
A range of different stainless steel accessories to use with Optima™ baths.
- Raised shelves help accommodate different sizes of vessel by altering the effective liquid depth
- Robust test tube racks are available in a variety of configurations for all sizes of tubes
Ordering information: ST5 and P5 baths can accommodate 1 of the following racks: 462-8251, 462-8252, 462-8253, 462-8254, 462-8255 and 462-8256, the ST12 or P12 and larger baths can accommodate 2 or more of the following racks 462-8257, 462-8258, 462-8133, 462-8134, 462-8261 or 462-8262.
Specifications
- Description:Raised shelf
- For:ST18/P18