Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OMEGA MEMBRANE 62MM 50K, Ultrafiltration Membrane, UF, ideal for use in stirred cell 1 * 12 items
Catalog # 516-8627
Supplier:
Cytiva (Formerly Pall Lab)
Expand image
OMEGA MEMBRANE 62MM 50K, Ultrafiltration Membrane, UF, ideal for use in stirred cell 1 * 12 items
Catalog # 516-8627
Supplier:
Cytiva (Formerly Pall Lab)