Order Entry
Export
Orders LinkContactUsLinkComponent
Pipette calibration software, PICASO

Pipette calibration software, PICASO

Vessel holder (1 set of 1 × Ø 15 mm and 1 × Ø 24 mm)
Catalog # EPPE3113806.000
   Low Manufacturing Impact
Supplier:   EPPENDORF
Pipette calibration software, PICASO
Catalog # EPPE3113806.000
Supplier:  EPPENDORF
   Low Manufacturing Impact
Pipette calibration software, PICASO
Pipette calibration software, PICASO

Vessel holder (1 set of 1 × Ø 15 mm and 1 × Ø 24 mm)

Price
46,60 €
Log in for availability
1 * 1 items
Product Details & Documents

PICASO (PipetteCalibrationSoftware) is a program for the automatic input and processing of measuring data from the gravimetric or photometric pipette test. With PICASO, all types of pipettes and other dispensers (e.g. bottle-top dispensers) can be checked.

  • User-oriented Windows interface, multilingual
  • On-line operating instructions and help function
  • Complete storage of a measurement including all individual values
  • Transfer of the data to other Windows programs (e.g. Excel) possible
  • Supports the archiving of complete data records
  • Analytical balances (Mettler (model AT and AE), Sartorius (model RC and MC), OHAUS (model AP) and AnD (model HM)can be connected via RS232 interface

System requirements:
IBM compatible computer in following minimum configuration:
– CPU: min. 80486/40Mhz under Windows 3.1 or 3.11 or min. Intel - Pentium/75Mhz under Windows 95
– RAM: 8 MB under 3.1 Windows or 3.11; 16 MB under Windows 95
– Hard disk (EIDE or SCSI) with at least 20 MB free disk space
– VGA monitor

Specifications
  • Description:
    Vessel holder (1 set of 1 × Ø 15 mm and 1 × Ø 24 mm)
  • Cat. No.:
    EPPE3113806.000
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact