Order Entry
Export
Orders LinkContactUsLinkComponent
PCR microtubes, 0,2 ml

PCR microtubes, 0,2 ml

PCR tube strips, 8-tube strips with individually attached caps (960 tubes), Clear, 0,2 ml
Catalog # 732-0098
   Low Manufacturing Impact
Supplier:   EPPENDORF
PCR microtubes, 0,2 ml
Catalog # 732-0098
Supplier:  EPPENDORF
   Low Manufacturing Impact
PCR microtubes, 0,2 ml
PCR microtubes, 0,2 ml

PCR tube strips, 8-tube strips with individually attached caps (960 tubes), Clear, 0,2 ml

Price
202,00 €
Log in for availability
1 * 120 items
Product Details & Documents

PP.

  • 0,2 ml tube has a contamination shield on the hinged lid
  • Certified free from DNase, RNase and human genomic DNA

These thin-walled, 0,2 ml PCR tubes are designed for use with all leading thermal cyclers.

Colour: Clear

Specifications
  • Capacity:
    0,2 ml
  • Colour:
    Clear
  • Description:
    PCR tube strips, 8-tube strips with individually attached caps (960 tubes)
  • Seq:
    02
  • Cat. No.:
    732-0098
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  • Catalog #
    Packed
    Availability
    Price
  • Frame, for work tray (set of 5)
    Frame, for work tray (set of 5)
    211-2111
    1 Set
    Log in for availability
    1 * 5 items - 58,80 €