Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Product Details & Documents
Eppendorf® Safe-Lock tubes 0,5 ml - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.
- Hinged Safe-Lock lid prevents unintended opening during incubation and storage
- g-Safe® technology, dependent on the Safe-Lock tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
- g-Safe® technology prevents sample loss in the event of tube breakage, and provides enhanced safety when handling hazardous samples
- Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures
- Precise lid sealing minimises evaporation rates during long-term storage
- Large frosted lid and side surface facilitate easy labeling
- Ensured functionality in a temperature range from −86 to +100 °C
- Autoclavable when open (121 °C, 20 min.) for effective sterilisation
- Available in lot-certified PCR clean, Eppendorf Forensic DNA grade, and Biopur® quality, ensuring tested purity
Eppendorf® Safe-Lock 0,5 ml microtubes epitomise advanced sample storage, featuring a patented Safe-Lock design for unparalleled security. Tailored for PCR applications, these microtubes provide a space-efficient 0,5 ml capacity, ideal for labs with limited storage. Their compatibility with various centrifuge models ensures seamless integration into existing workflows. Robust construction and user-friendly attributes, such as clear graduations, further enhance reliability and ease of handling. Elevate laboratory practices with Eppendorf® Safe-Lock tubes, which set a new standard for sample integrity and operational efficiency.
Manufactured without the use of slip agents, plasticisers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Cap type: Safe-Lock
Material: PP
Volume: 0,5 ml
Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.
Specifications
- Packed:500 tubes ( 5 bags×100 tubes)
- Type:With attached lid
- Version:Graduated, conical bottom, non sterile
- Capacity:0,5 ml
- Colour:Assorted
- Seq:07
- Cat. No.:211-2149
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Temperature range:−86…+100 °C
- Autoclavable:When open 121 °C, 20 mins
- Environmentally Preferable:Low Manufacturing Impact



