Order Entry
Export
Orders LinkContactUsLinkComponent
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes

Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes

0,5 ml, Red, With attached lid, Graduated, conical bottom, non sterile, 500 tubes (1 bag×500 tubes)
Catalog # 211-2142
   Low Manufacturing Impact
Supplier:   EPPENDORF
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Catalog # 211-2142
Supplier:  EPPENDORF
   Low Manufacturing Impact
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes

0,5 ml, Red, With attached lid, Graduated, conical bottom, non sterile, 500 tubes (1 bag×500 tubes)

Price
68,90 €
Log in for availability
1 * 500 items
Product Details & Documents

Eppendorf® Safe-Lock tubes 0,5 ml - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

  • Hinged Safe-Lock lid prevents unintended opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss in the event of tube breakage, and provides enhanced safety when handling hazardous samples
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures
  • Precise lid sealing minimises evaporation rates during long-term storage
  • Large frosted lid and side surface facilitate easy labeling
  • Ensured functionality in a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for effective sterilisation
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA grade, and Biopur® quality, ensuring tested purity

Eppendorf® Safe-Lock 0,5 ml microtubes epitomise advanced sample storage, featuring a patented Safe-Lock design for unparalleled security. Tailored for PCR applications, these microtubes provide a space-efficient 0,5 ml capacity, ideal for labs with limited storage. Their compatibility with various centrifuge models ensures seamless integration into existing workflows. Robust construction and user-friendly attributes, such as clear graduations, further enhance reliability and ease of handling. Elevate laboratory practices with Eppendorf® Safe-Lock tubes, which set a new standard for sample integrity and operational efficiency.

Manufactured without the use of slip agents, plasticisers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Cap type: Safe-Lock
Material: PP
Volume: 0,5 ml

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Specifications
  • Packed:
    500 tubes (1 bag×500 tubes)
  • Type:
    With attached lid
  • Version:
    Graduated, conical bottom, non sterile
  • Capacity:
    0,5 ml
  • Colour:
    Red
  • Seq:
    04
  • Cat. No.:
    211-2142
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Temperature range:
    −86…+100 °C
  • Autoclavable:
    When open 121 °C, 20 mins
  • Environmentally Preferable:
    Low Manufacturing Impact