Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Sieves
Test Sieves
Accessories for Test Sieves
Accessories for Test Sieves
Print
Accessories for Test Sieves
Collector, 200 mm, Stainless steel
Catalog # 510-0273
Supplier:
ENDECOTTS
Accessories for Test Sieves
Catalog # 510-0273
Supplier:
ENDECOTTS
Expand image
Return to Top
Accessories for Test Sieves
Collector, 200 mm, Stainless steel
Specifications
Add to cart
Jump to:
Specifications
Price
112,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Ø:
200 mm
Description:
Collector
Material:
Stainless steel
Seq:
05