Orders LinkContactUsLinkComponent
Accessories for Test Sieves
Intermediate collector, 200 mm, Stainless steel
Catalog # ENDE200S/STINT
Supplier:   ENDECOTTS
Accessories for Test Sieves
Catalog # ENDE200S/STINT
Supplier:  ENDECOTTS
Accessories for Test Sieves
Intermediate collector, 200 mm, Stainless steel
Price
151,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Ø:
    200 mm
  • Description:
    Intermediate collector
  • Material:
    Stainless steel
  • Seq:
    09