Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
BUFFER SOLUTION PH 4.001 @ 20°C NIST 1 * 500 mL
BUFFER SOLUTION PH 4.001 @ 20°C NIST 1 * 500 mL
Print
BUFFER SOLUTION PH 4.001 @ 20°C NIST 1 * 500 mL
Catalog # REGE104001
Certificates
Supplier:
REAGECON
BUFFER SOLUTION PH 4.001 @ 20°C NIST 1 * 500 mL
Catalog # REGE104001
Supplier:
REAGECON
Certificates
Expand image
Return to Top
BUFFER SOLUTION PH 4.001 @ 20°C NIST 1 * 500 mL
Add to cart
Price
41,70 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number