Bodies for Stemi 2000
Body with apochromatic Greenough system and stereoscopic angle 11°, 0,65× - 5,0×
Catalog # 630-0370
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Body with apochromatic Greenough system and stereoscopic angle 11°, 0,65× - 5,0×
- Seq:01