Bodies for Stemi 2000
Body with apochromatic Greenough with stereoscopic angle 11°, 0,65 - 5,0× and dust protection cap with M49×0,75 for photo filter
Catalog # 630-0367
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Body with apochromatic Greenough with stereoscopic angle 11°, 0,65 - 5,0× and dust protection cap with M49×0,75 for photo filter
- Seq:02