Orders LinkContactUsLinkComponent
Lenses for Stemi 2000
Lenses for Stemi 2000
Front lens, 210 mm
Catalog # 630-0361
Supplier:   ZEISS
Lenses for Stemi 2000
Catalog # 630-0361
Supplier:  ZEISS
Lenses for Stemi 2000
Lenses for Stemi 2000
Front lens, 210 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    0,40×
  • Description:
    Front lens
  • Seq:
    002
  • Working distance:
    210 mm