Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Condenser 0.55 (a=31mm) for objective magnifications 5x-40x(VAREL contrast 10x...40x) 1 * 1 items
Catalog # 630-1918
Supplier:
ZEISS
Expand image
Condenser 0.55 (a=31mm) for objective magnifications 5x-40x(VAREL contrast 10x...40x) 1 * 1 items
Catalog # 630-1918
Supplier:
ZEISS