Order Entry
Export
Orders LinkContactUsLinkComponent
 

ISE standard solutions

Standard solution calcium ions (ES/Ca), 1 L
  662-0368
 :   WTW
ISE standard solutions
  662-0368
 :  WTW
 :  
ISE standard solutions
ISE standard solutions

Standard solution calcium ions (ES/Ca), 1 L

 :  
 
164,00 €
   
1 * 1 L
 

These standard solutions for calibration contain 10 g of the ion to be measured per 10 liters.

 

 

 

  • Pk:
    1 L
  • Description:
    Standard solution calcium ions (ES/Ca)