Order Entry
Export
Orders LinkContactUsLinkComponent
 

ISE standard solutions

Standard solution chloride (ES/Cl), 1 L
Catalog # 662-1420
Supplier:   WTW
ISE standard solutions
Catalog # 662-1420
Supplier:  WTW
CAS Number:  
ISE standard solutions
ISE standard solutions

Standard solution chloride (ES/Cl), 1 L

CAS Number:  
Price
164,00 €
Log in for availability
1 * 1 L
Product Details & Documents

These standard solutions for calibration contain 10 g of the ion to be measured per 10 liters.

 

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Standard solution chloride (ES/Cl)

Specifications