Buffer solutions as per DIN 19266-01: 2000
Buffer solution, PL 12, 12,47, ±0,02, 250 mL
Supplier: WTW
Jump to:
Product Details & Documents
These standard buffer solutions (work reference buffer solutions) are widely used in precision applications.
- Traceably certified to PTB/NIST and therefore safe for use
- Every bottle is labeled with their own lot number with an expiration date
Certifications: DIN 19266-01: 2000.
Documents
Certificates
Search by Lot Numbercoc_WTWA109000_07.27 CoC_WTWA109000_08.24-1 CoC_WTWA109000_12.23-1 CoC_WTWA109110_01.24-1 CoC_WTWA109110_02.27-1
More
Sensors for Field and Laboratory.pdf
Additional Product Details
WTWA109110_TDS_EN
Datasheet
Specifications
Specifications
- Pk:250 mL
- Description:Buffer solution, PL 12
- pH accuracy:±0,02
- pH value:12,47



