Orders LinkContactUsLinkComponent
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
Universal sensor extension assembly UA130 (incl. handle and set of seals), for mounting and protecting all 650, 690, 70X and 70X IQ series sensors, length without sensor: 1067 mm
Catalog # 662-0246
Supplier:   WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Catalog # 662-0246
Supplier:  WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
Universal sensor extension assembly UA130 (incl. handle and set of seals), for mounting and protecting all 650, 690, 70X and 70X IQ series sensors, length without sensor: 1067 mm
Price
288,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Universal sensor extension assembly UA130 (incl. handle and set of seals), for mounting and protecting all 650, 690, 70X and 70X IQ series sensors, length without sensor: 1067 mm
  • Seq:
    27