Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for pH electrodes
Accessories for pH electrodes
Print
Accessories for pH electrodes
Cable, 1 m, with S7/BNC plug
Catalog # 662-1327
Supplier:
WTW
Accessories for pH electrodes
Catalog # 662-1327
Supplier:
WTW
Expand image
Return to Top
Accessories for pH electrodes
Cable, 1 m, with S7/BNC plug
Specifications
Add to cart
Jump to:
Specifications
Price
59,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Additional Product Details
Sensors for Field and Laboratory.pdf
Specifications
Description:
Cable, 1 m, with S7/BNC plug
Seq:
02