Orders LinkContactUsLinkComponent
Vial racks, WHEATON®
Vial racks, WHEATON®
30 mm, 50, 337×170×30 mm
Catalog # 548-1976
Supplier:   DWK Life Sciences
Vial racks, WHEATON®
Catalog # 548-1976
Supplier:  DWK Life Sciences
Vial racks, WHEATON®
Vial racks, WHEATON®
30 mm, 50, 337×170×30 mm
Price
550,00 €
Log in for availability
1 * 5 items
Product Details & Documents
These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
  • Autoclavable and stackable
  • Good chemical resistance
  • Includes position identifiers
Specifications
  • Hole Ø:
    30 mm
  • No. of Holes:
    50
  • Seq:
    02
  • W×D×H:
    337×170×30 mm