Vial racks, WHEATON®
15,5 mm, 48, 266×94×28 mm
Catalog # 548-2157
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
- Autoclavable and stackable
- Good chemical resistance
- Includes position identifiers
Specifications
- Hole Ø:15,5 mm
- No. of Holes:48
- Seq:01
- W×D×H:266×94×28 mm



