Trypsinising flasks, with screw caps, WHEATON®
700 ml, Top screw cap 51-400
WHEA355397
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
- Capacity:700 ml
- Description:Top screw cap 51-400