Orders LinkContactUsLinkComponent
The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates an d the other for filling a 384-well plate. For increased productivity and minimal plastic waste, the use of PIPETMAN® 1 * 1 items
The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates an d the other for filling a 384-well plate. For increased productivity and minimal plastic waste, the use of PIPETMAN® 1 * 1 items
Catalog # GILSFH07001
New Product
Supplier:   GILSON
The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates an d the other for filling a 384-well plate. For increased productivity and minimal plastic waste, the use of PIPETMAN® 1 * 1 items
Catalog # GILSFH07001
Supplier:  GILSON
New Product
The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates an d the other for filling a 384-well plate. For increased productivity and minimal plastic waste, the use of PIPETMAN® 1 * 1 items
Price
550,00 €
Log in for availability
1 * 1 items
Product Details & Documents