Conductivity Standards
Product Details & Documents
To enable analysts to achieve the correct result when measuring conductivity and to prove the correctness of that result; Reagecon has developed a high quality range of Conductivity Standards; which can be used for calibration of conductivity sensors; quality control; method validation and instrument qualification.
Reagecon Conductivity Standards are aqueous based; thereby eliminating any errors attributable to matrix mismatch
Directly traceable to NIST Standard Reference Materials; meets the specification requirements of all the major Pharmacopoeias
The low temperature coefficient of variation; reduces measurement errors and enables non-temperature controlled use and is suitable for field work
High accuracy ±1% at 25 °C; enables the Standards to be used as calibrators and/or controls; in accordance with the most exacting industrial requirements
Manufactured to exacting specifications with an extended shelf life and stability; even after opening the bottle thereby eliminating the requirement to open a fresh bottle of Standard every time it is used
Tested at 25 °C
Used with conductivity meters
Packaging: HDPE Bottle GL 45
Physical Form: Liquid
Specifications
Specifications
- Description:Conductivity reference standard
- Conductivity:50 µS/cm ± 1%



