Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
[1,1'-BI(CYCLOHEXAN)]-2-OL,98%,5G 1 * 5 g
[1,1'-BI(CYCLOHEXAN)]-2-OL,98%,5G 1 * 5 g
Print
[1,1'-BI(CYCLOHEXAN)]-2-OL,98%,5G 1 * 5 g
Catalog # BLDPBD74216-5G
New Product
Supplier:
BLD PHARMATECH
[1,1'-BI(CYCLOHEXAN)]-2-OL,98%,5G 1 * 5 g
Catalog # BLDPBD74216-5G
Supplier:
BLD PHARMATECH
New Product
CAS Number:
Expand image
Return to Top
[1,1'-BI(CYCLOHEXAN)]-2-OL,98%,5G 1 * 5 g
Add to cart
CAS Number:
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 5 g
-
+
Inquire For Price
Return Policy
Product Details & Documents