Orders LinkContactUsLinkComponent
Pressure Bulbs
Pressure Bulbs
Ball set with net, merlot-red, size 5, natural rubber/PVC, 102
Catalog # DENE4120003
New Product
Supplier:   DEUTSCH NEUMANN
Pressure Bulbs
Catalog # DENE4120003
Supplier:  DEUTSCH NEUMANN
New Product
Pressure Bulbs
Pressure Bulbs
Ball set with net, merlot-red, size 5, natural rubber/PVC, 102
Price
108,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Specifications
  • Capacity:
    102
  • Description:
    Ball set with net, merlot-red, size 5, natural rubber/PVC
  • Seq:
    5