Order Entry
Export
Orders LinkContactUsLinkComponent
Conductivity Standards

Conductivity Standards

Conductivity reference standard, 1000 µS/cm ± 1.0%
Catalog # REGECRMCSKC1000
New Product
Supplier:   REAGECON
Conductivity Standards
Catalog # REGECRMCSKC1000
Supplier:  REAGECON
New Product
CAS Number:  
Conductivity Standards

Conductivity reference standard, 1000 µS/cm ± 1.0%

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

To enable analysts to achieve the correct result when measuring conductivity and to prove the correctness of that result; Reagecon has developed a high quality range of Conductivity Standards; which can be used for calibration of conductivity sensors; quality control; method validation and instrument qualification.

  • Reagecon Conductivity Standards are aqueous based; thereby eliminating any errors attributable to matrix mismatch

    Directly traceable to NIST Standard Reference Materials; meets the specification requirements of all the major Pharmacopoeias

    The low temperature coefficient of variation; reduces measurement errors and enables non-temperature controlled use and is suitable for field work

    High accuracy ±1% at 25 °C; enables the Standards to be used as calibrators and/or controls; in accordance with the most exacting industrial requirements

    Manufactured to exacting specifications with an extended shelf life and stability; even after opening the bottle thereby eliminating the requirement to open a fresh bottle of Standard every time it is used

Tested at 25 °C

Used with conductivity meters

Packaging: HDPE Bottle GL 45

Physical Form: Liquid

Specifications

Specifications

  • Description:
    Conductivity reference standard
  • Conductivity:
    1000 µS/cm ± 1.0%

Specifications