Micro Powder Spatulas, Angular/Semi-Cup-Shaped
Product Details & Documents
The micro powder spatula is made of corrosion-resistant stainless steel 1.4301 (304) and is specially designed for precise dosing and contamination-free work in analytical laboratories. Its slim shape and dual-sided design allow for precise handling of small powder quantities, even in narrow containers such as vials or test tubes.
Two functions: flat side for spreading, semicircular side for picking up and transporting
Slim shaft enables work in narrow vessels such as vials, test tubes and bottles
Fine blade for precise dosing and pinpoint accuracy when working with very small quantities
The polished surface prevents residue buildup and makes cleaning easy. This spatula is an indispensable tool for applications requiring precision and hygiene, such as sample preparation, weighing sensitive substances, and spreading on analysis surfaces.
The micro powder spatula is ideal for use in chemical, pharmaceutical, and analytical laboratories, where precise and safe processing of small quantities of powder is necessary. Typical applications include preparing samples for spectroscopic analysis, weighing sensitive substances, and working in narrow vessels. The spatula is also used in environmental, food, and material analysis.
Specifications
- Colour:Silver
- Description:Micro powder spatula, angular/semi-cup-shaped
- Length:150 mm
- Seq:1