Order Entry
Export
Orders LinkContactUsLinkComponent
Test Tubes with Screw Cap

Test Tubes with Screw Cap

10 ml, Yes, Screw cap, Test tubes, sterile individually wrapped
Catalog # KART5604
New Product
Supplier:   KARTELL
Test Tubes with Screw Cap
Catalog # KART5604
Supplier:  KARTELL
New Product
Test Tubes with Screw Cap

10 ml, Yes, Screw cap, Test tubes, sterile individually wrapped

Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents

PP test tubes, 10 ml, with self-standing base and tight screw cap yellow colour for a leakproof closure.

  • Not graduated

    Available also in sterile and individually wrapped version

    Ø×H: 16×100 mm

Specifications
  • Cap included:
    Yes
  • Closure type:
    Screw cap
  • Capacity:
    10 ml
  • Description:
    Test tubes, sterile individually wrapped
  • Seq:
    2