Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) nitrate 99+%

Catalog # 211560000.
Irritant
Health
Environment
Oxidizing
Corrosive
New Product
Lead(II) nitrate 99+%
Catalog # 211560000.
New Product
CAS Number:  10099-74-8
Lead(II) nitrate 99+%
Lead(II) nitrate 99+%
CAS Number:  10099-74-8
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:13,05432
  • Melting Pt:470 °C (dec.)
  • Density:4.53 g/cm³
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II

 

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Titration Complexometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Iron (Fe):
    ≤10 ppm