Trypan Blue 80%
Bottle
Catalog # APOSOR10154002.5KG
New Product
Supplier: Apollo Scientific
Trypan Blue 80%
Catalog # APOSOR10154002.5KG
Supplier: Apollo Scientific
New Product
CAS Number: 72-57-1
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
- CAS Number:72-57-1
- Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL Number:MFCD00003969
- Molecular Weight:960,81 g/mol
Specifications
Specifications
- Pack type:Bottle



