Order Entry
Export
Orders LinkContactUsLinkComponent
Cetyldimethylethylammonium Bromide 99%

Cetyldimethylethylammonium Bromide 99%

Bottle
Catalog # APOSOR10126352.5KG
Irritant
Environment
New Product
Supplier:   Apollo Scientific
Cetyldimethylethylammonium Bromide 99%
Catalog # APOSOR10126352.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  124-03-8
Cetyldimethylethylammonium Bromide 99%

Bottle

CAS Number:  124-03-8
Price
414,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/reagent

  • Store at 2 to 8 °C
Synonyms:  Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide
  • CAS Number:124-03-8
  • Formula:C₂₀H₄₄BrN
  • MDL Number:MFCD00011774
  • Molecular Weight:378,47 g/mol
  • Storage Temperature:Refrigerator
  • UN:3077
Specifications

Specifications

  • Pack type:
    Bottle

Specifications