Order Entry
Export
Orders LinkContactUsLinkComponent
Ac-Tyr(Tbu)-OH 98%

Ac-Tyr(Tbu)-OH 98%

Bottle
Catalog # APOSOR1012216250MG
New Product
Supplier:   Apollo Scientific
Ac-Tyr(Tbu)-OH 98%
Catalog # APOSOR1012216250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  201292-99-1
Ac-Tyr(Tbu)-OH 98%

Bottle

CAS Number:  201292-99-1
Price
21,80 €
Log in for availability
1 * 250 mg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Ac-Tyr(tBu)-OH
  • CAS Number:201292-99-1
  • Formula:C₁₅H₂₁NO₄
  • MDL Number:MFCD24532698
  • Molecular Weight:279,33 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications