Order Entry
Export
Orders LinkContactUsLinkComponent
(E)-2-Bromocinnamaldehyde 96%

(E)-2-Bromocinnamaldehyde 96%

Bottle
Catalog # APOSOR1008335-1G
New Product
Supplier:   Apollo Scientific
(E)-2-Bromocinnamaldehyde 96%
Catalog # APOSOR1008335-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  138555-58-5
(E)-2-Bromocinnamaldehyde 96%

Bottle

CAS Number:  138555-58-5
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:138555-58-5
  • Formula:C₉H₇BrO
  • MDL Number:MFCD03032509
  • Molecular Weight:211,06 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications