0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
n-Hexacosane
N-Hexacosane 99%
N-Hexacosane 99%
Bottle
APOSOR1014386-500G
New Product
:
Apollo Scientific
N-Hexacosane 99%
APOSOR1014386-500G
:
Apollo Scientific
New Product
:
630-01-3
N-Hexacosane 99%
Bottle
:
630-01-3
1 960,00 €
1 * 500 g
-
+
Usage: Organic building block/reagent
:
N-hexacosane
:
630-01-3
:
C₂₆H₅₄
:
MFCD00009354
:
366,71 g/mol
Pack type:
Bottle