Order Entry
Export
Orders LinkContactUsLinkComponent
N-Hexacosane 99%

N-Hexacosane 99%

Bottle
Catalog # APOSOR1014386-1G
New Product
Supplier:   Apollo Scientific
N-Hexacosane 99%
Catalog # APOSOR1014386-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  630-01-3
N-Hexacosane 99%

Bottle

CAS Number:  630-01-3
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  N-hexacosane
  • CAS Number:630-01-3
  • Formula:C₂₆H₅₄
  • MDL Number:MFCD00009354
  • Molecular Weight:366,71 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications