0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Eosin B
BP0000697420
Eosin B
Bottle
APOSOR1013107-25G
New Product
:
Apollo Scientific
Eosin B
APOSOR1013107-25G
:
Apollo Scientific
New Product
:
Eosin B
Bottle
:
24,60 €
1 * 25 g
-
+
Pack type:
Bottle