0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
(±)-10-Hydroxycamptothecin
BP0000725851
10-Hydroxycamptothecin
Bottle
APOSOR1009313-25G
New Product
:
Apollo Scientific
10-Hydroxycamptothecin
APOSOR1009313-25G
:
Apollo Scientific
New Product
:
10-Hydroxycamptothecin
Bottle
:
646,00 €
1 * 25 g
-
+
Pack type:
Bottle