Order Entry
Export
Orders LinkContactUsLinkComponent
2-Mercaptopyrimidine 98%

2-Mercaptopyrimidine 98%

Bottle
Catalog # APOSOR1010512-5G
New Product
Supplier:   Apollo Scientific
2-Mercaptopyrimidine 98%
Catalog # APOSOR1010512-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  131242-36-9
2-Mercaptopyrimidine 98%

Bottle

CAS Number:  131242-36-9
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  2-Thiopyrimidine
  • CAS Number:131242-36-9
  • Formula:C₄H₄N₂S
  • MDL Number:MFCD00006073
  • Molecular Weight:112,15 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications