0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,9-Decadiene
BP0000690069
1,9-Decadiene
Bottle
APOSOR10092992.5KG
New Product
:
Apollo Scientific
1,9-Decadiene
APOSOR10092992.5KG
:
Apollo Scientific
New Product
:
1,9-Decadiene
Bottle
:
3 410,00 €
1 * 2,5 kg
-
+
Pack type:
Bottle