Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,9-Decadiene 98%

Bottle
Catalog # APOSOR1009299-25G
New Product
Supplier:   Apollo Scientific
1,9-Decadiene 98%
Catalog # APOSOR1009299-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  1647-16-1
1,9-Decadiene 98%

Bottle

CAS Number:  1647-16-1
Price
52,10 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Deca-1,9-diene
  • CAS Number:1647-16-1
  • Formula:C₁₀H₁₈
  • MDL Number:MFCD00008670
  • Molecular Weight:138,25 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications