0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,2'-O-Cyclouridine
BP0000708093
2,2'-O-Cyclouridine
Bottle
APOSOR1009951-5G
New Product
:
Apollo Scientific
2,2'-O-Cyclouridine
APOSOR1009951-5G
:
Apollo Scientific
New Product
:
2,2'-O-Cyclouridine
Bottle
:
21,80 €
1 * 5 g
-
+
Pack type:
Bottle