0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,1-Diethoxycyclohexane
BP0000707989
1,1-Diethoxycyclohexane
1,1-Diethoxycyclohexane
Bottle
APOSOR1009042-5G
New Product
:
Apollo Scientific
1,1-Diethoxycyclohexane
APOSOR1009042-5G
:
Apollo Scientific
New Product
:
Expand image
1,1-Diethoxycyclohexane
Bottle
Add to cart
:
46,40 €
Unit of Measure
1 * 5 g
-
+
Add to cart
Pack type:
Bottle