Order Entry
Export
Orders LinkContactUsLinkComponent
1-Naphthohydrazide 98%

1-Naphthohydrazide 98%

Bottle
Catalog # APOSOR1009547-500G
New Product
Supplier:   Apollo Scientific
1-Naphthohydrazide 98%
Catalog # APOSOR1009547-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  43038-45-5
1-Naphthohydrazide 98%

Bottle

CAS Number:  43038-45-5
Price
1 670,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  1-Naphthalenecarbohydrazide
  • CAS Number:43038-45-5
  • Formula:C₁₁H₁₀N₂O
  • MDL Number:MFCD19613794
  • Molecular Weight:186,21 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications