1,2-Bis(2,3,4,5,6-Pentabromophenyl)Ethane 98%
Bottle
Catalog # APOSOR1009085-25KG
New Product
Supplier: Apollo Scientific
1,2-Bis(2,3,4,5,6-Pentabromophenyl)Ethane 98%
Catalog # APOSOR1009085-25KG
Supplier: Apollo Scientific
New Product
CAS Number: 84852-53-9
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: 2,2',3,3',4,4',5,5',6,6'-Decabromobibenzyl, 1,2-Bis(perbromophenyl)ethane
- CAS Number:84852-53-9
- Formula:C₁₄H₄Br₁₀
- MDL Number:MFCD06407713
- Molecular Weight:971,22 g/mol
Specifications
Specifications
- Pack type:Bottle



